Learn More
Cole-Parmer Oakton by Cole-Parmer ISE Electrode, Ammonia, Loose Membrane Cap; BNC
Catalog No. 50633669 Shop All Cole-Parmer Products
Encompass_Preferred
Easy-to-use, single-junction electrodes feature durable, epoxy bodies.The electrode design lessens errors and provides accurate results within 30 seconds, depending on solution concentration. Concentration range is 0.01 to 17,000 ppm. Use it over a 0 to 50 degree Celsius temperature range and above pH 11. The BNC connector and one-meter cable allow you to use with almost any existing meter.
Catalog No. 50-633-669 Supplier Cole-Parmer Supplier No. 2750200
Easy-to-use, single-junction electrodes feature durable, epoxy bodies.The electrode design lessens errors and provides accurate results within 30 seconds, depending on solution concentration. Concentration range is 0.01 to 17,000 ppm. Use it over a 0 to 50 degree Celsius temperature range and above pH 11. The BNC connector and one-meter cable allow you to use with almost any existing meter.