Learn More
Mettler Toledo™ Electrode Arm, EasyPlace Complete
Catalog No. 01911931 Shop All METTLER TOLEDO Products
Change view
Click to view available options
For Use With (Equipment):
EasyPlace
Catalog No. For Use With (Equipment)
01-911-931 EasyPlace
1 options
Catalog No. 01-911-931 Supplier Mettler Toledo™ Supplier No. 30671569
Add to Cart
Add to Cart
The EasyPlace electrode arm supports efficient measurement and calibration. With perfect vertical movement, spillage risks are minimized. Includes parking beaker, parking beaker insert and sachet holder.
  • Precise Control
  • Perfectly vertical sensor movement makes it easy to insert the sensor into samples
  • The right place
  • The EasyPlace contains dedicated positions for calibration, short- and long term sensor storage and measurement, simplifying everyday workflows
  • Fits right in
  • The EasyPlace electrode arm fits together with the EasyMix magnetic stirrer and standard sachets, simplifying operation and saving space
TRUSTED_SUSTAINABILITY
Specifications
Type Electrode Arm, EasyPlace Complete
For Use With (Equipment) EasyPlace