Learn More
Cole-Parmer Oakton by Cole-Parmer Combination Ion-Selective Electrode (ISE), Bromide (Br)
Catalog No. 50633625 Shop All Cole-Parmer Products
Encompass_Preferred
Easily fit this electrode of bromid with epoxy body into small spaces without any added reference electrode.
Catalog No. 50-633-625 Supplier Cole-Parmer Supplier No. 2750402
Easily fit this electrode of bromid with epoxy body into small spaces without any added reference electrode.