Learn More
Cole-Parmer Oakton by Cole-Parmer Ion-Selective Electrode (ISE), Calcium, Double-Junction, BNC
Catalog No. 50633631 Shop All Cole-Parmer Products
Encompass_Preferred
Easily fit this electrode of calcium with glass body into small spaces without any added reference electrode.
Catalog No. 50-633-631 Supplier Cole-Parmer Supplier No. 2750209
Easily fit this electrode of calcium with glass body into small spaces without any added reference electrode.