Learn More
Hach Company Hach RENOVO.X Electrode Cleaning Solution, Extra Strong, 250 mL (S16M002)
Catalog No. NC2046440 Shop All Hach Company Products
Encompass
Hach RENOVO.X Electrode Cleaning Solution (Extra Strong) is a 250 mL cleaning reagent formulated to remove stubborn protein and organic deposits from pH electrodes and sensors. It is intended for maintenance cleaning after exposure to dirty water or biologically active samples, helping restore electrode response and extend sensor lifetime.
  • Removes difficult protein and organic deposits.
  • Designed for pH electrodes and sensors.
  • Helps restore electrode performance and extend sensor lifetime.
  • Convenient 250 mL bottle suitable for lab and field maintenance.
  • Sodium hypochlorite-based formula for effective protein cleaning.
Catalog No. NC2046440 Supplier Hach Company Supplier No. S16M002
Hach RENOVO.X Electrode Cleaning Solution (Extra Strong) is a 250 mL cleaning reagent formulated to remove stubborn protein and organic deposits from pH electrodes and sensors. It is intended for maintenance cleaning after exposure to dirty water or biologically active samples, helping restore electrode response and extend sensor lifetime.
  • Removes difficult protein and organic deposits.
  • Designed for pH electrodes and sensors.
  • Helps restore electrode performance and extend sensor lifetime.
  • Convenient 250 mL bottle suitable for lab and field maintenance.
  • Sodium hypochlorite-based formula for effective protein cleaning.