Learn More
Hach Company Electrode Cleaning Solution for Regular Maintenance, 500 mL
Catalog No. 501996960 Shop All Hach Company Products
Encompass
Electrode cleaning solution for pH electrode regular cleaning maintenance. Convenient for soft cleaning of the pH sensing element after use in clean water samples. 500 mL bottle.
Catalog No. 50-199-6960 Supplier Hach Company Supplier No. 2965249
Electrode cleaning solution for pH electrode regular cleaning maintenance. Convenient for soft cleaning of the pH sensing element after use in clean water samples. 500 mL bottle.