Learn More
Hach Company Electrode Cleaning Solution
Catalog No. 5000206977 Shop All Hach Company Products
Encompass
Electrode Cleaning Solution for Proteins/Organics Samples-250 mLElectrode cleaning solution to remove proteins residues organic contaminations/deposits such as food stuff from pH sensing element 250 mL Bottle
Catalog No. 50-002-06977 Supplier Hach Company Supplier No. C20C370
Electrode Cleaning Solution for Proteins/Organics Samples-250 mLElectrode cleaning solution to remove proteins residues organic contaminations/deposits such as food stuff from pH sensing element 250 mL Bottle