Learn More
Hach Company Electrode Cleaning Solution
Catalog No. 5000207224 Shop All Hach Company Products
Encompass
Electrode Cleaning Solution for Porous Pin/Diaphragm Junction-250 mLElectrode cleaning solution for combined pH/Metal porous pin/diaphragm cleaning from Sulphides or AgCl precipitate/deposit/clogging
Catalog No. 50-002-07224 Supplier Hach Company Supplier No. C20C380
Electrode Cleaning Solution for Porous Pin/Diaphragm Junction-250 mLElectrode cleaning solution for combined pH/Metal porous pin/diaphragm cleaning from Sulphides or AgCl precipitate/deposit/clogging