Learn More
Hach Company Radiometer Analytical PHC3359-8 Combination pH Electrode for Microsamples (d=3 mm, glass body, BNC)
Catalog No. 501997587 Shop All Hach Company Products
Encompass
The Radiometer PHC3359-8 Combination pH Electrode has a glass body, Ag/AgCl reference system, refillable electrolyte (3M KCl + sat. AgCl) and micro diameter (3 mm) for small volume sample measurements. The PHC3359-8 has a BNC connector and is compatible with Hach Sension+ laboratory pH meters.
Catalog No. 50-199-7587 Supplier Hach Company Supplier No. E16M343
The Radiometer PHC3359-8 Combination pH Electrode has a glass body, Ag/AgCl reference system, refillable electrolyte (3M KCl + sat. AgCl) and micro diameter (3 mm) for small volume sample measurements. The PHC3359-8 has a BNC connector and is compatible with Hach Sension+ laboratory pH meters.