Learn More
Hach Company Radiometer Analytical PHC2002-8 Combination Red-Rod pH Electrode with long length (length=200 mm, glass body, BNC)
Catalog No. 501997578 Shop All Hach Company Products
Encompass
The Radiometer PHC2002-8 Combination pH Electrode has a glass body, Red-Rod reference system, refillable electrolyte (sat. KCl) and porous pin junction. The PHC2002-8 has a long length of 200 mm for general purpose applications. The PHC2002-8 is compatible with Hach Sension+ laboratory pH meters (or any meter with a BNC connection).
Catalog No. 50-199-7578 Supplier Hach Company Supplier No. E16M315
The Radiometer PHC2002-8 Combination pH Electrode has a glass body, Red-Rod reference system, refillable electrolyte (sat. KCl) and porous pin junction. The PHC2002-8 has a long length of 200 mm for general purpose applications. The PHC2002-8 is compatible with Hach Sension+ laboratory pH meters (or any meter with a BNC connection).