Learn More
Hach Company Radiometer Analytical PHC2005-8 Combination Red-Rod pH Electrode (epoxy body, BNC)
Catalog No. 501997589 Shop All Hach Company Products
Encompass
The Radiometer PHC2005-8 Combination pH Electrode has a robust epoxy body, Red-Rod reference system, refillable electrolyte (sat. KCl) and porous pin junction for general purpose applications. The PHC2005-8 is compatible with Hach Sension+ laboratory pH meters (or any meter with a BNC connection).
Catalog No. 50-199-7589 Supplier Hach Company Supplier No. E16M500
The Radiometer PHC2005-8 Combination pH Electrode has a robust epoxy body, Red-Rod reference system, refillable electrolyte (sat. KCl) and porous pin junction for general purpose applications. The PHC2005-8 is compatible with Hach Sension+ laboratory pH meters (or any meter with a BNC connection).