Learn More
Hach Company Radiometer Analytical PHC2011-8 Combination Red-Rod pH Electrode for Alkaline Samples (alkaline glass, glass body, BNC)
Catalog No. 501997579 Shop All Hach Company Products
Encompass
The Radiometer PHC2011-8 Combination pH Electrode has a glass body, Red-Rod reference system, refillable electrolyte (sat. KCl) and porous pin junction. The PHC2011-8 has a glass membrane specific for alkaline samples. The PHC2011-8 is compatible with Hach Sension+ laboratory pH meters (or any meter with a BNC connection).
Catalog No. 50-199-7579 Supplier Hach Company Supplier No. E16M317
The Radiometer PHC2011-8 Combination pH Electrode has a glass body, Red-Rod reference system, refillable electrolyte (sat. KCl) and porous pin junction. The PHC2011-8 has a glass membrane specific for alkaline samples. The PHC2011-8 is compatible with Hach Sension+ laboratory pH meters (or any meter with a BNC connection).