Learn More
Hach Company Radiometer Analytical PHC2051-8 Combination Red-Rod pH Electrode with robust glass sensor (glass body, BNC)
Catalog No. 501997581 Shop All Hach Company Products
Encompass
The Radiometer PHC2051-8 Combination pH Electrode has a glass body, Red-Rod reference system, refillable electrolyte (sat. KCl) and porous pin junction. The PHC2051-8 has a special thick glass membrane for measurement in soils or other applications requiring a robust sensor. The PHC2051-8 is compatible with Hach Sension+ laboratory pH meters (or any meter with a BNC connection).
Catalog No. 50-199-7581 Supplier Hach Company Supplier No. E16M319
The Radiometer PHC2051-8 Combination pH Electrode has a glass body, Red-Rod reference system, refillable electrolyte (sat. KCl) and porous pin junction. The PHC2051-8 has a special thick glass membrane for measurement in soils or other applications requiring a robust sensor. The PHC2051-8 is compatible with Hach Sension+ laboratory pH meters (or any meter with a BNC connection).