Learn More
Hach Company Radiometer Analytical PHC2085-8 Combination Red-Rod pH Electrode with Temperature Sensor (epoxy body, BNC+cinch)
Catalog No. 501997590 Shop All Hach Company Products
Encompass
The Radiometer PHC2085-8 Combination pH Electrode has a robust epoxy body, Red-Rod reference system, refillable electrolyte (sat. KCl), integrated temperature sensor, and porous pin junction for general purpose applications. The PHC2085-8 has BNC + cinch (RCA) connectors. Please reference the PHC2085-6 (article no. E16M345) for a version of this probe compatible with Hach Sension+ laboratory pH meters.
Catalog No. 50-199-7590 Supplier Hach Company Supplier No. E16M501
The Radiometer PHC2085-8 Combination pH Electrode has a robust epoxy body, Red-Rod reference system, refillable electrolyte (sat. KCl), integrated temperature sensor, and porous pin junction for general purpose applications. The PHC2085-8 has BNC + cinch (RCA) connectors. Please reference the PHC2085-6 (article no. E16M345) for a version of this probe compatible with Hach Sension+ laboratory pH meters.