Learn More
Hach Company Radiometer Analytical PHC2401-8 Combination Red-Rod pH Electrode (glass body, BNC)
Catalog No. 501997588 Shop All Hach Company Products
Encompass
The Radiometer PHC2401-8 Combination pH Electrode has a glass body, Red-Rod reference system, refillable electrolyte (sat. KCl) and annular ring junction for general purpose applications. The PHC2401-8 is compatible with Hach Sension+ laboratory pH meters (or any meter with a BNC connection).
Catalog No. 50-199-7588 Supplier Hach Company Supplier No. E16M400
The Radiometer PHC2401-8 Combination pH Electrode has a glass body, Red-Rod reference system, refillable electrolyte (sat. KCl) and annular ring junction for general purpose applications. The PHC2401-8 is compatible with Hach Sension+ laboratory pH meters (or any meter with a BNC connection).