Learn More
Hach Company Radiometer Analytical PHC2701-8 Combination Red-Rod pH Electrode (high-flow junction, glass body, BNC)
Catalog No. 501997583 Shop All Hach Company Products
Encompass
The Radiometer PHC2701-8 Combination pH Electrode has a glass body, Red-Rod reference system, refillable electrolyte (sat. KCl) and high-flow annular ring junction. The high-flow junction is ideal for low-ionic strength samples, dairy, or other challenging applications. The PHC2701-8 is compatible with Hach Sension+ laboratory pH meters (or any meter with a BNC connection).
Catalog No. 50-199-7583 Supplier Hach Company Supplier No. E16M323
The Radiometer PHC2701-8 Combination pH Electrode has a glass body, Red-Rod reference system, refillable electrolyte (sat. KCl) and high-flow annular ring junction. The high-flow junction is ideal for low-ionic strength samples, dairy, or other challenging applications. The PHC2701-8 is compatible with Hach Sension+ laboratory pH meters (or any meter with a BNC connection).