Learn More
Hach Company Radiometer Analytical PHC3081-8 Combination pH Electrode with Temperature Sensor (glass body, Ag/AgCl ref., BNC+cinch)
Catalog No. 501997576 Shop All Hach Company Products
Encompass
The Radiometer PHC3081-8 Combination pH Electrode has a glass body, Ag/AgCl reference system, refillable electrolyte (3M KCl + sat. AgCl), integrated temperature sensor, and porous pin junction for general purpose applications. The PHC3081-8 has BNC + cinch (RCA) connectors. Please reference the PHC3081-6 (article no. E16M344) for a version of this probe compatible with Hach Sension+ laboratory pH meters.
Catalog No. 50-199-7576 Supplier Hach Company Supplier No. E16M305
The Radiometer PHC3081-8 Combination pH Electrode has a glass body, Ag/AgCl reference system, refillable electrolyte (3M KCl + sat. AgCl), integrated temperature sensor, and porous pin junction for general purpose applications. The PHC3081-8 has BNC + cinch (RCA) connectors. Please reference the PHC3081-6 (article no. E16M344) for a version of this probe compatible with Hach Sension+ laboratory pH meters.