Learn More
Hach Company Radiometer Analytical PHC3185-8 Combination pH Electrode with Temperature Sensor (gel electrolyte, epoxy body, BNC+cinch)
Catalog No. 501997584 Shop All Hach Company Products
Encompass
The Radiometer PHC3185-8 Combination pH Electrode has a robust epoxy body, Ag/AgCl reference system, non-refillable gel electrolyte (3M KCl), integrated temperature sensor, and open junction for general purpose applications. The PHC3185-8 has BNC + cinch (RCA) connectors. Please note that the PHC3185-8 is not compatible with Hach Sension+ laboratory pH meters.
Catalog No. 50-199-7584 Supplier Hach Company Supplier No. E16M329
The Radiometer PHC3185-8 Combination pH Electrode has a robust epoxy body, Ag/AgCl reference system, non-refillable gel electrolyte (3M KCl), integrated temperature sensor, and open junction for general purpose applications. The PHC3185-8 has BNC + cinch (RCA) connectors. Please note that the PHC3185-8 is not compatible with Hach Sension+ laboratory pH meters.