Learn More
Med Vet International Ecg Monitoring Electrode Tape Backing Non-Radiolucent Snap Connector, 2239, 50/Bag
Catalog No. 502829533
Encompass_Preferred
Micropore™ Tape Backing backing Without abrader A round-style monitoring electrode Micropore Tape Backing backing is breathable, flexible and conformable, yet is gentle to the skin for increased patient comfort Patented solid gel conforms to the skin quickly allowing high quality, uninterrupted traces The gel's low chloride content minimizes skin irritation for increased patient comfort3M™ SureSeal Cap Seal prevents premature product dry out to help ensure product freshness for 30 days out of the bag
Catalog No. 50-282-9533 Supplier Med Vet International Supplier No. 187583BG
Micropore™ Tape Backing backing Without abrader A round-style monitoring electrode Micropore Tape Backing backing is breathable, flexible and conformable, yet is gentle to the skin for increased patient comfort Patented solid gel conforms to the skin quickly allowing high quality, uninterrupted traces The gel's low chloride content minimizes skin irritation for increased patient comfort3M™ SureSeal Cap Seal prevents premature product dry out to help ensure product freshness for 30 days out of the bag