Learn More
Med Vet International Ecg Monitoring Electrode Foam Backing Non-Radiolucent Snap Connector, 2560, 1000/Case
Catalog No. 502832310
Encompass_Preferred
Without -purpose monitoring electrodes with Foam Backing backing and sticky gel are ideally suited for the OR, ER and diagnostic stress testing applications Conveniently packaged in individual strips of ten electrodes for easy use and less waste Remains fresh for 30 days out of the bag High performance adhesive offers instant adhesion and up to five-day wear with excellent patient skin care and comfort Sticky gel delivers excellent trace quality resulting in better patient information
Catalog No. 50-283-2310 Supplier Med Vet International Supplier No. 368360CS
Without -purpose monitoring electrodes with Foam Backing backing and sticky gel are ideally suited for the OR, ER and diagnostic stress testing applications Conveniently packaged in individual strips of ten electrodes for easy use and less waste Remains fresh for 30 days out of the bag High performance adhesive offers instant adhesion and up to five-day wear with excellent patient skin care and comfort Sticky gel delivers excellent trace quality resulting in better patient information