Learn More
Cole-Parmer Epoxy-Body pH Electrode with ATC, Large Ceramic Junction, Gel-Filled; 1/EA
Catalog No. 50633531 Shop All Cole-Parmer Products
Encompass_Preferred
The Ag/AgCl combination reference electrodes have an impact-resistant epoxy body. Electrodes provide a fast, stable response and are ideal for prolonged readings. Gel-filled design requires virtually no maintenance. 127 mm L x 12 mm diameter with maximum 176F (80C) temperature.
Catalog No. 50-633-531 Supplier Cole-Parmer Supplier No. 0599214
The Ag/AgCl combination reference electrodes have an impact-resistant epoxy body. Electrodes provide a fast, stable response and are ideal for prolonged readings. Gel-filled design requires virtually no maintenance. 127 mm L x 12 mm diameter with maximum 176F (80C) temperature.