Learn More
Cole-Parmer Oakton Basic conductivity/tds meter
Catalog No. 50633530 Shop All Cole-Parmer Products
Encompass_Preferred
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.
Catalog No. 50-633-530 Supplier Cole-Parmer Supplier No. 0599216
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.