Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Siemens Healthineers Electrode Fill Solutions

Catalog No. 23044868 Shop All Siemens Healthineers Products
Change view
Click to view available options
Electrode Fill Solution:
NA+/K+/CA++/CL-
pH
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Electrode Fill Solution
23-044-868 NA+/K+/CA++/CL-
23-044-867 pH
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-044-868 Supplier Siemens Healthineers Supplier No. 10376882
Add to Cart
Add to Cart
This item is not returnable. View return policy

In vitro diagnostics material for use with RAPIDLab™ 1200 series electrodes.

TRUSTED_SUSTAINABILITY

Specifications

Electrode Fill Solution NA+/K+/CA++/CL-
For Use With (Equipment) RAPIDLab™ 1200 Series Electrodes
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.