Learn More
Med Vet International Ecg Monitoring Electrode Foam Backing Radiolucent / Mr Conditional Snap Connector, 2570, 50/Bag
Catalog No. 502832052
Encompass_Preferred
Built in Abrader Multi-purpose monitoring electrodes with Foam Backing backing and sticky gel are ideally suited for the OR, ER and diagnostic stress testing applications Conveniently packaged in individual strips of ten electrodes for easy use and less waste Remains fresh for 30 days out of the bag High performance adhesive offers instant adhesion and up to five-day wear with excellent patient skin care and comfort Sticky gel delivers excellent trace quality resulting in better patient information Micro replicated Trace Prep surface gently prepares skin for optimal trace quality Radiolucent
Catalog No. 50-283-2052 Supplier Med Vet International Supplier No. 368361BG
 Restricted Item
Add to Cart
Add to Cart
Built in Abrader Multi-purpose monitoring electrodes with Foam Backing backing and sticky gel are ideally suited for the OR, ER and diagnostic stress testing applications Conveniently packaged in individual strips of ten electrodes for easy use and less waste Remains fresh for 30 days out of the bag High performance adhesive offers instant adhesion and up to five-day wear with excellent patient skin care and comfort Sticky gel delivers excellent trace quality resulting in better patient information Micro replicated Trace Prep surface gently prepares skin for optimal trace quality Radiolucent