Learn More
World Precision Instrument Electrode Handle 4.0 mm
SDP
Catalog No. 502521252
Encompass
Electrode holder for popular manipulators
This 4.0 mm electrode holder is designed for use with Narishige and Zeiss manipulators
Catalog No. 50-252-1252 Supplier World Precision Instrument Supplier No. 5444
Electrode holder for popular manipulators
This 4.0 mm electrode holder is designed for use with Narishige and Zeiss manipulators