Learn More
Cole-Parmer Cole-Parmer Electrode Holder for Removable pH and ORP Electrodes, CPVC
Catalog No. 50633437 Shop All Cole-Parmer Products
Encompass_Preferred
Electrode holders have threads on each end. Install holders into a pipe tee for in-line use or on the end of a submersion pipe. The integral cap assembly permits quick servicing for calibrations or replacement.Choose from CPVC for economical aplications or PVDF for enhanced chemical resistance. Electrodes order separately.
Catalog No. 50-633-437 Supplier Cole-Parmer Supplier No. 2700950
Electrode holders have threads on each end. Install holders into a pipe tee for in-line use or on the end of a submersion pipe. The integral cap assembly permits quick servicing for calibrations or replacement.Choose from CPVC for economical aplications or PVDF for enhanced chemical resistance. Electrodes order separately.