Learn More
World Precision Instrument Electrode Holder for KWIK-Tips
SDP
Catalog No. 502521156
Encompass
  • Use with KWIK-TIP Ion Selective Electrodes
  • TIPA-Calcium
  • TIPH-Hydrogen
  • TIPK-Potassium
  • TIPTPP-TPP+
Catalog No. 50-252-1156 Supplier World Precision Instrument Supplier No. KWIK2
  • Use with KWIK-TIP Ion Selective Electrodes
  • TIPA-Calcium
  • TIPH-Hydrogen
  • TIPK-Potassium
  • TIPTPP-TPP+