Learn More
METTLER TOLEDO pH Sensor InPro 3250i SG/225
Catalog No. NC0109829 Shop All METTLER TOLEDO Products
Encompass
The InPro 3250i SG/225 is a pre-pressurized, liquid filled, low maintenance pH & ORP sensor, combined with a temperature sensor, for in-line measurement in demanding applications. The 225 mm version is optimal for medium installation lengths.
Catalog No. NC0109829 Supplier METTLER TOLEDO Supplier No. 52005374
The InPro 3250i SG/225 is a pre-pressurized, liquid filled, low maintenance pH & ORP sensor, combined with a temperature sensor, for in-line measurement in demanding applications. The 225 mm version is optimal for medium installation lengths.