Learn More
Cole-Parmer Cole-Parmer ELECTRODE ION-SELECT BROMIDE
Supplier:  Cole-Parmer 3581204
Encompass_Preferred
These industrial sealed double-junction electrodes with stripped wire ends easily connect to the ion selective controller. Industrial electrodes will withstand the conditions of your in-line application. Electrodes measure 12 mm dia x 80 mm L with 10-ft cord.
Catalog No. 50-196-1682