Learn More
Cole-Parmer Oakton by Cole-Parmer Ion-Selective Electrode (ISE), Lead, Double-Junction, BNC
Catalog No. 50633680 Shop All Cole-Parmer Products
Encompass_Preferred
Easily fit this electrode of lead with glass body into small spaces without any added reference electrode.
Catalog No. 50-633-680 Supplier Cole-Parmer Supplier No. 2750225
Easily fit this electrode of lead with glass body into small spaces without any added reference electrode.