Learn More
Cole-Parmer Cole-Parmer Removable Double-Junction ORP Electrode
Catalog No. 50633492 Shop All Cole-Parmer Products
Encompass_Preferred
Use for oxidation/reduction measurements in plating baths, waste-water monitoring, and other general-purpose applications. Platinum band ORP withstands temperatures from 23 to 212ºF (-5 to 100ºC).
Catalog No. 50-633-492 Supplier Cole-Parmer Supplier No. 2700931
Use for oxidation/reduction measurements in plating baths, waste-water monitoring, and other general-purpose applications. Platinum band ORP withstands temperatures from 23 to 212ºF (-5 to 100ºC).