Learn More
Cole-Parmer Cole-Parmer Double-Junction ORP Electrode, Gold Sensor, 10-ft Cable
Catalog No. 50633582 Shop All Cole-Parmer Products
Encompass_Preferred
ORP electrodes measure oxidation/reduction levels in plating baths, waste water monitoring, and more. Use platinum-band electrodes for general-purpose applications from 23 to 212°>F (-5 to 100°>C). Use gold-disc electrodes for solutions containing cyanide from 23 to 230°>F (-5 to 110°>C).
Catalog No. 50-633-582 Supplier Cole-Parmer Supplier No. 2700625
ORP electrodes measure oxidation/reduction levels in plating baths, waste water monitoring, and more. Use platinum-band electrodes for general-purpose applications from 23 to 212°>F (-5 to 100°>C). Use gold-disc electrodes for solutions containing cyanide from 23 to 230°>F (-5 to 110°>C).