Learn More
Cole-Parmer Oakton ORP electrode; double-junction, BNC Connector
Catalog No. 501961392 Shop All Cole-Parmer Products
Encompass_Preferred
The Ag/AgCl combination reference electrodes have an impact-resistant epoxy body. Electrodes provide a fast, stable response and are ideal for prolonged readings. Gel-filled design requires virtually no maintenance. 127 mm L x 12 mm diameter with maximum 176F (80C) temperature.
Catalog No. 50-196-1392 Supplier Cole-Parmer Supplier No. 5900177
The Ag/AgCl combination reference electrodes have an impact-resistant epoxy body. Electrodes provide a fast, stable response and are ideal for prolonged readings. Gel-filled design requires virtually no maintenance. 127 mm L x 12 mm diameter with maximum 176F (80C) temperature.