Learn More
Cole-Parmer Oakton by Cole-Parmer Self-Cleaning Flat Surface ORP Probe, DJ/Ryton/Pt/10'/BNC
Catalog No. 50633489 Shop All Cole-Parmer Products
Encompass_Preferred
The flat surface of this electrode prevents buildup of solids.
Catalog No. 50-633-489 Supplier Cole-Parmer Supplier No. 2700350
The flat surface of this electrode prevents buildup of solids.