Learn More
Cole-Parmer Cole-Parmer Solution Grounded, Platinum-Band ORP Electrode; No ATC Element, BNC
Catalog No. 501961234 Shop All Cole-Parmer Products
Encompass_Preferred
These electrodes offer solution ground, PTFE references with double junctions and polymerized gel. All models include a 10-ft cable with BNC pH connector and Ryton housing with 3/4'' NPT(M) threads.
Catalog No. 50-196-1234 Supplier Cole-Parmer Supplier No. 2701840
These electrodes offer solution ground, PTFE references with double junctions and polymerized gel. All models include a 10-ft cable with BNC pH connector and Ryton housing with 3/4'' NPT(M) threads.