Learn More
Cole-Parmer Cole-Parmer Tuff-Tip pH Electrode 100 Ohm PT RTD(Mini Audio Plug), BNC, 3/4"
Catalog No. 50633697 Shop All Cole-Parmer Products
Encompass_Preferred
Don't worry about bulb breakage, the unique Tuff-Tip design helps protect bulbs. Choose chemical-resistant Ryton PPS standard electrode or with graphite tip. Eliminate electrical noise with graphite-tipped models. Ideal for viscous or severe applications, these electrodes reduce contamination and clogging with uniform pore size.
Catalog No. 50-633-697 Supplier Cole-Parmer Supplier No. 2702002
Don't worry about bulb breakage, the unique Tuff-Tip design helps protect bulbs. Choose chemical-resistant Ryton PPS standard electrode or with graphite tip. Eliminate electrical noise with graphite-tipped models. Ideal for viscous or severe applications, these electrodes reduce contamination and clogging with uniform pore size.