Learn More
Cole-Parmer Cole-Parmer General Purpose pH Electrode, 10-ft Cable
Supplier:  Cole-Parmer 0599420
Encompass_Preferred
General-purpose single-junction electrodes are economical yet accurate. Can be used for measurements up to pH 13. Maximum pressure is 150 psi at 25°>C.
Catalog No. 50-633-563