Learn More
Cole-Parmer Cole-Parmer General Purpose pH electrode, double-junction, 10-ft cable
Catalog No. 50633569 Shop All Cole-Parmer Products
Encompass_Preferred
This general purpose, double-junction electrode prevents contamination of the AgCl reference cell by solutions that might react with silver, such as heavy metals, organics and sulfides.
Catalog No. 50-633-569 Supplier Cole-Parmer Supplier No. 0599422
This general purpose, double-junction electrode prevents contamination of the AgCl reference cell by solutions that might react with silver, such as heavy metals, organics and sulfides.