Learn More
Cole-Parmer Cole-Parmer pH Probe, DJ/Glass/100Ohm RTD/12mm/10'; Tinned/BNC
Catalog No. 50633566 Shop All Cole-Parmer Products
Encompass_Preferred
General purpose, double-junction electrodes prevent contamination of the AgCl reference cell by solutions that might react with silver (heavy metals, organics, sulfides). Electrodes can be used for measurements up to pH 13. Maximum pressure is 150 psi at 25°C.
Catalog No. 50-633-566 Supplier Cole-Parmer Supplier No. 2701300
General purpose, double-junction electrodes prevent contamination of the AgCl reference cell by solutions that might react with silver (heavy metals, organics, sulfides). Electrodes can be used for measurements up to pH 13. Maximum pressure is 150 psi at 25°C.