Learn More
Cole-Parmer Cole-Parmer pH Probe, SJ/Glass/100Ohm RTD/12 mm/10'; Tinned/BNC
Catalog No. 50633565 Shop All Cole-Parmer Products
Encompass_Preferred
General-purpose single-junction electrodes are economical yet accurate. Can be used for measurements up to pH 13. Maximum pressure is 150 psi at 25°>C.
Catalog No. 50-633-565 Supplier Cole-Parmer Supplier No. 0599401
General-purpose single-junction electrodes are economical yet accurate. Can be used for measurements up to pH 13. Maximum pressure is 150 psi at 25°>C.