Learn More
Cole-Parmer Cole-Parmer Tuff-Tip, Semi-Dome 1" In-Line/Sub pH, 100Ohm tinned ATC
Catalog No. 50633696 Shop All Cole-Parmer Products
Encompass_Preferred
Don't worry about bulb breakage— unique design helps protect bulbs – Eliminate electrical noise with graphite-tipped models – Ideal for viscous or severe applications – Reduce contamination and clogging with uniform pore size
Catalog No. 50-633-696 Supplier Cole-Parmer Supplier No. 2702023
Don't worry about bulb breakage— unique design helps protect bulbs – Eliminate electrical noise with graphite-tipped models – Ideal for viscous or severe applications – Reduce contamination and clogging with uniform pore size