Learn More
Cole-Parmer Cole-Parmer Tuff-Tip pH Electrode, 3/4" 100 Ohm RTD
Catalog No. 50633699 Shop All Cole-Parmer Products
Encompass_Preferred
Don't worry about bulb breakage because the unique Tuff-Tip design helps to protect bulbs. There are chemical-resistant Ryton PPS standard electrodes or ones with graphite tips, which eliminate electrical noise. They are ideal for viscous or severe applications and reduce contamination and clogging with uniform pore size.
Catalog No. 50-633-699 Supplier Cole-Parmer Supplier No. 2702043
Don't worry about bulb breakage because the unique Tuff-Tip design helps to protect bulbs. There are chemical-resistant Ryton PPS standard electrodes or ones with graphite tips, which eliminate electrical noise. They are ideal for viscous or severe applications and reduce contamination and clogging with uniform pore size.