Learn More
Cole-Parmer Cole-Parmer Removable Blue Glass Double-Junction pH Electrode
Catalog No. 50633490 Shop All Cole-Parmer Products
Encompass_Preferred
Double-junction electrodes prevent electrode contamination by solutions that might react with Ag+ or Cl- (heavy metals, organics, and sulfides). Use for measurements up to 13 pH. Withstands temperatures form 23 to 212F (-5 to 100C).
Catalog No. 50-633-490 Supplier Cole-Parmer Supplier No. 2700910
Double-junction electrodes prevent electrode contamination by solutions that might react with Ag+ or Cl- (heavy metals, organics, and sulfides). Use for measurements up to 13 pH. Withstands temperatures form 23 to 212F (-5 to 100C).