Learn More
Cole-Parmer Cole-Parmer pH electrode, combination, rugged, epoxy body, interchangeable connection
Catalog No. 50633520 Shop All Cole-Parmer Products
Encompass_Preferred
Match these combination electrodes with one of the connector cables listed at the right. If you use several pH meters that each have different connector types, these electrodes and cables can save you money. The probe-cable interface is moisture proof and noise free, with a sealing O-ring, gold-plated contacts, and PTFE insulation.
Catalog No. 50-633-520 Supplier Cole-Parmer Supplier No. 0565845
Add to Cart
Add to Cart
Match these combination electrodes with one of the connector cables listed at the right. If you use several pH meters that each have different connector types, these electrodes and cables can save you money. The probe-cable interface is moisture proof and noise free, with a sealing O-ring, gold-plated contacts, and PTFE insulation.