Learn More
Cole-Parmer Cole-Parmer pH Electrode for Ultrapure Water
Catalog No. 50633433 Shop All Cole-Parmer Products
Encompass_Preferred
Electrode has 10-ft long cable with BNC pH connector and tinned end for solution ground.
Catalog No. 50-633-433 Supplier Cole-Parmer Supplier No. 2700450
Electrode has 10-ft long cable with BNC pH connector and tinned end for solution ground.