Learn More
Cole-Parmer Oakton by Cole-Parmer self-cleaning pH electrode, PPS housing, 100 Ohms Pt RTD
Catalog No. 50633460 Shop All Cole-Parmer Products
Encompass_Preferred
There will be no further buildups with this self-cleaning electrode cable. Its flat surface prevents buildups, while fluid flow cleans surface for longer life. It can be used with most meters and controllers. The BNC connector and ATC element work with a variety of units. Its double junction is for difficult applications.
Catalog No. 50-633-460 Supplier Cole-Parmer Supplier No. 2700323
There will be no further buildups with this self-cleaning electrode cable. Its flat surface prevents buildups, while fluid flow cleans surface for longer life. It can be used with most meters and controllers. The BNC connector and ATC element work with a variety of units. Its double junction is for difficult applications.