Learn More
Cole-Parmer Cole-Parmer Interchangeable Cable Refillable SJ Glass pH Probe; 329 x 8 mm
Catalog No. 50633512 Shop All Cole-Parmer Products
Encompass_Preferred
Match these combination electrodes with one of the connector cables listed at the right. If you use several pH meters that each have different connector types, these electrodes and cables can save you money. The probe-cable interface is moisture proof and noise free, with a sealing O-ring, gold-plated contacts, and PTFE insulation.
Catalog No. 50-633-512 Supplier Cole-Parmer Supplier No. 0565835
Match these combination electrodes with one of the connector cables listed at the right. If you use several pH meters that each have different connector types, these electrodes and cables can save you money. The probe-cable interface is moisture proof and noise free, with a sealing O-ring, gold-plated contacts, and PTFE insulation.