Learn More
Cole-Parmer Oakton by Cole-Parmer pH electrode, combination, double-junction, KNO3 gel, glass body, BNC
Catalog No. 50633567 Shop All Cole-Parmer Products
Encompass_Preferred
Glass-body combination electrodes feature refillable references, except for 05998-30 which features a high-temperature KNO3 gel. All have BNC connectors.
Catalog No. 50-633-567 Supplier Cole-Parmer Supplier No. 0599830
Glass-body combination electrodes feature refillable references, except for 05998-30 which features a high-temperature KNO3 gel. All have BNC connectors.