Learn More
Cole-Parmer Cole-Parmer pH Probe, Refillable, High-temp, pH with high Na+
Catalog No. 50633574 Shop All Cole-Parmer Products
Encompass_Preferred
Glass body combination probes are able to withstand higher temperatures and chemical attack. All have 3.3 ft (1 m) long cables with BNC connectors.
Catalog No. 50-633-574 Supplier Cole-Parmer Supplier No. 0599132
Glass body combination probes are able to withstand higher temperatures and chemical attack. All have 3.3 ft (1 m) long cables with BNC connectors.