Learn More
Cole-Parmer Cole-Parmer pH electrode, Refillable, low maintenance
Catalog No. 50633505 Shop All Cole-Parmer Products
Encompass_Preferred
This electrode gives a faster response and increased stability across a pH range of 0 to 14. A dome bulb offers better protection from breakage. It has stability in low-ionic strength solutions and offers resistance to Na+ ion interference. It is compatible with most brands of pH meters that accept standard BNC connections.
Catalog No. 50-633-505 Supplier Cole-Parmer Supplier No. 0599181
This electrode gives a faster response and increased stability across a pH range of 0 to 14. A dome bulb offers better protection from breakage. It has stability in low-ionic strength solutions and offers resistance to Na+ ion interference. It is compatible with most brands of pH meters that accept standard BNC connections.